Anti-PUS7 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX102422
89318-746EA 582.76 USD
89318-746
Anti-PUS7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Collagen IV alpha2 (collagen, type IV, alpha 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PUS7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR