Anti-QPRT Mouse Monoclonal Antibody [clone: MF4]

Supplier: Genetex

GTX10239
89336-816EA 358 USD
89336-816
Anti-QPRT Mouse Monoclonal Antibody [clone: MF4]
Antibodies
Mouse monoclonal [MF4] to cardiac Troponin I

This antibody is recommend for better sensitivity in WB. <br> <br>Recommended pairs for ELISA: <br>19C7 (GTX28289) + M18 (GTX10236) mixture for capture and MF4 (this antibody) + 16A11 (GTX10226) mixture for detection <br>M18 (GTX10236) + 560 (GTX38795) mixture for capture and MF4 (this antibody) + 19C7 (GTX28289) mixture for detection <br>MF4 (this antibody) + 560 (GTX38795) mixture for capture and 19C7 (GTX28289) + 267 (GTX14530) mixture for detection

Type: Primary
Antigen: QPRT
Clonality: Monoclonal
Clone: MF4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR