Anti-RABL2A Mouse Monoclonal Antibody [clone: M155]

Supplier: Genetex

GTX10237
89336-814EA 358 USD
89336-814
Anti-RABL2A Mouse Monoclonal Antibody [clone: M155]
Antibodies
Mouse monoclonal [M155] to cardiac Troponin I

Type: Primary
Antigen: RABL2A
Clonality: Monoclonal
Clone: M155
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Dog, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR