Anti-RALBP1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX102365
89335-354EA 582.76 USD
89335-354
Anti-RALBP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to DP2 (transcription factor Dp-2 (E2F dimerization partner 2))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RALBP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR