Anti-RALBP1 Mouse Monoclonal Antibody [clone: M18]

Supplier: Genetex

GTX10236
89336-812EA 358 USD
89336-812
Anti-RALBP1 Mouse Monoclonal Antibody [clone: M18]
Antibodies
Mouse monoclonal [M18] to cardiac Troponin I

Recommended pairs for ELISA: <br>19C7 (GTX28289) + M18 (this antibody) mixture for capture and MF4 (GTX10239) + 16A11 (GTX10226) mixture for detection <br>M18 (this antibody) + 560 (GTX38759) mixture for capture and MF4 (GTX10239) + 19C7 (GTX28289) mixture for detection <br>M18 (GTX10236) + 560 (GTX38759) mixture for capture and 19C7 (GTX28289) + 267 (GTX14530) mixture for detection

Type: Primary
Antigen: RALBP1
Clonality: Monoclonal
Clone: M18
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR