Anti-RASD2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX102341
89308-304EA 176.03 USD
89308-304
Anti-RASD2 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to MFGE8 (milk fat globule-EGF factor 8 protein)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: RASD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR