Anti-RNF144B Mouse Monoclonal Antibody [clone: 3C7]

Supplier: Genetex

GTX10230
89336-808EA 358 USD
89336-808
Anti-RNF144B Mouse Monoclonal Antibody [clone: 3C7]
Antibodies
Mouse monoclonal [3C7] to cardiac Troponin I

Type: Primary
Antigen: RNF144B
Clonality: Monoclonal
Clone: 3C7
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR