Anti-RNF144B Rabbit Polyclonal Antibody

Supplier: Genetex

GTX102298
89308-292EA 404.77 USD
89308-292
Anti-RNF144B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to IGFBP4 (insulin-like growth factor binding protein 4)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: RNF144B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR