Anti-RNF144B Rabbit Polyclonal Antibody

Supplier: Genetex

GTX102296
89318-696EA 394.91 USD
89318-696
Anti-RNF144B Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Cpn10 (heat shock 10kDa protein 1 (chaperonin 10))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RNF144B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR