Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RNF144B Mouse Monoclonal Antibody [clone: 23C6]

Supplier: Genetex

GTX10229
89336-806EA 325.28 USD InStock
89336-806
Anti-RNF144B Mouse Monoclonal Antibody [clone: 23C6]
Antibodies
Mouse monoclonal [23C6] to cardiac Troponin I

Type: Primary
Antigen: RNF144B
Clonality: Monoclonal
Clone: 23C6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty