Anti-RNF144B Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX102272
89308-284EA 176.03 USD
89308-284
Anti-RNF144B Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to SREBP-1 (sterol regulatory element binding transcription factor 1)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: RNF144B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR