Anti-RPA2 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX102264
89318-692EA 394.91 USD
89318-692
Anti-RPA2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to KIF1C (kinesin family member 1C)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RPA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR