Anti-RPA2 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX102258
89308-272EA 793.5 USD
89308-272
Anti-RPA2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PRA1 (Rab acceptor 1 (prenylated))

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: RPA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR