Anti-SATB1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX102204
89318-682EA 582.76 USD
89318-682
Anti-SATB1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SAR1A (SAR1 homolog A (S. cerevisiae))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SATB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR