Anti-SCYL3 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX102198
89318-678EA 427.1 USD
89318-678
Anti-SCYL3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to MEK2 (mitogen-activated protein kinase kinase 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SCYL3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR