Anti-SDR9C7 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX102190
89318-670EA 582.76 USD
89318-670
Anti-SDR9C7 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SUMO2/3 (SMT3 suppressor of mif two 3 homolog 2 (S. cerevisiae))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SDR9C7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR