Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SDR9C7 Mouse Monoclonal Antibody [clone: 5C12]

Supplier: Genetex

GTX10219
89336-802EA 325.28 USD InStock
89336-802
Anti-SDR9C7 Mouse Monoclonal Antibody [clone: 5C12]
Antibodies
Mouse monoclonal [5C12] to cardiac Troponin T

AP: Use at an assay dependent dilution. ELISA: Use at an assay dependent dilution. IA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 35.4 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SDR9C7
Clonality: Monoclonal
Clone: 5C12
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty