Anti-SIL1 Mouse Monoclonal Antibody [clone: 12A6]

Supplier: Genetex

GTX10209
89336-514EA 271.29 USD
89336-514
Anti-SIL1 Mouse Monoclonal Antibody [clone: 12A6]
Antibodies
Mouse monoclonal [12A6] to Transferrin

ELISA: Use at an assay dependent dilution. Can be used for the detection of transferrin in serum by sandwich ELISA as an antibody pair with GTX10212 as capture antibody and GTX10209 for detection. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SIL1
Clonality: Monoclonal
Clone: 12A6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR