Anti-SIL1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX102083
89308-236EA 394.91 USD
89308-236
Anti-SIL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to hnRNP H (heterogeneous nuclear ribonucleoprotein H1 (H))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SIL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR