Anti-SLC2A5 Mouse Monoclonal Antibody [clone: TA40]

Supplier: GeneTex
GTX10206
89336-510EA 759 USD
89336-510
Anti-SLC2A5 Mouse Monoclonal Antibody [clone: TA40]
Antibodies
Mouse monoclonal [TA40] to TNF alpha

FuncS: Use at an assay dependent dilution. Inhibits the biological activity of TNF alpha in different biological activities. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SLC2A5
Clonality: Monoclonal
Clone: TA40
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR