Anti-SLFNL1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX102023
89308-220EA 176.03 USD
89308-220
Anti-SLFNL1 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to GPR161 (G protein-coupled receptor 161)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: SLFNL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR