Anti-SOX17 Rabbit Polyclonal Antibody

GTX101933
89308-194EA 176.03 USD
89308-194
Anti-SOX17 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Exostosin-1 (exostoses (multiple) 1)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: SOX17
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR