Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SOX17 Mouse Monoclonal Antibody [clone: 10A5]

Supplier: Genetex

GTX10193
89336-498EA 246.49 USD InStock
89336-498
Anti-SOX17 Mouse Monoclonal Antibody [clone: 10A5]
Antibodies
Mouse monoclonal [10A5] to PAPP A

ELISA: Use at an assay dependent dilution. Can be used in immunoassay for the detection of human PAPPA antigen as an antibody pair with GTX10194 Mouse monoclonal [3C8] to PAPPA for coating. IA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Detects a band of approximately 500 kDa. Recognizes heterotetrameric complex in Western Blotting after SDS-electrophoresis in non-reducing conditions. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SOX17
Clonality: Monoclonal
Clone: 10A5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty