Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

Anti-SPR Mouse Monoclonal Antibody [clone: 4G11]

Supplier: Genetex

GTX10192
89336-496EA 308.16 USD InStock
89336-496
Anti-SPR Mouse Monoclonal Antibody [clone: 4G11]
Antibodies
Mouse monoclonal [4G11] to PAPP A

ELISA: Use at an assay dependent dilution. Can be paired for ELISA with GTX10194 Mouse monoclonal [3C8] to PAPPA and GTX10195 Mouse monoclonal [10H9] to PAPPA . WB: Use at an assay dependent dilution. Detects a band of approximately 500 kDa. GTX10192 recognizes heterotetrameric complex in Western Blotting after SDS-electrophoresis in non-reducing conditions. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SPR
Clonality: Monoclonal
Clone: 4G11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Monkey

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty