Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SRR Mouse Monoclonal Antibody [clone: PS2]

Supplier: Genetex
GTX10189
89336-492
Anti-SRR Mouse Monoclonal Antibody [clone: PS2]
Mouse monoclonal [PS2] to PSA

ELISA: Use at an assay dependent dilution. For the detection of PSA antigen use in ELISA as an antibody pair with GTX10190 ( GTX10189 coating and GTX10190 as conjugate). For the detection of free PSA use in ELISA as an antibody pair with GTX10188 (using either antibody to coat or as conjugate). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SRR
Clonality: Monoclonal
Clone: PS2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty