Anti-STAT5A Rabbit Polyclonal Antibody

Supplier: Genetex

GTX101846
89318-566EA 404.77 USD
89318-566
Anti-STAT5A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SNRPD2 (small nuclear ribonucleoprotein D2 polypeptide 16.5kDa)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:2000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: STAT5A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR