Anti-TBXAS1 Goat Polyclonal Antibody

Supplier: Genetex

GTX10180
89261-502EA 479.61 USD
89261-502
Anti-TBXAS1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to TCP1

Approx 60kDa band observed in Hela, Human Lung and Human Testis lysates (predicted MW of 60kDa according to NP_110379.2). Recommended for use at 0.5-2µg/ml. In paraffin embedded Human Testis shows only cytoplasmic staining in some germ cells, and both cytoplasmic and nuclear staining in other germ cells with unstained nucleoli. Recommended concentration, 1-2µg/ml. This antibody is expected to recognise both reported isoforms.

Type: Primary
Antigen: TBXAS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR