Anti-TBXAS1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101799
89318-538EA 582.76 USD
89318-538
Anti-TBXAS1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ATP6V1E1 (ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E1)

  • Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TBXAS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR