Anti-TPMT Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101740
89335-344EA 582.76 USD
89335-344
Anti-TPMT Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RASSF7 (Ras association (RalGDS/AF-6) domain family (N-terminal) member 7)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TPMT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR