Anti-TRIM33 Mouse Monoclonal Antibody [clone: 5H3]

Supplier: GeneTex
GTX10173
89317-172EA 449 USD
89317-172
Anti-TRIM33 Mouse Monoclonal Antibody [clone: 5H3]
Mouse monoclonal [5H3] to Plasminogen

AP: Use at an assay dependent dilution. ELISA: Use at an assay dependent dilution. IHC-P: Use at an assay dependent dilution. Antigen retrieval is not essential but may optimise staining. Can be used in ELISA with GTX10176 (conjugate). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TRIM33
Clonality: Monoclonal
Clone: 5H3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR