Anti-TRIM33 Rabbit Polyclonal Antibody

GTX101729
89318-514EA 376.43 USD
89318-514
Anti-TRIM33 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to G protein alpha Inhibitor 2 (guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TRIM33
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR