Anti-TTC32 Mouse Monoclonal Antibody [clone: P421]

GTX10168
89336-490EA 258.81 USD
89336-490
Anti-TTC32 Mouse Monoclonal Antibody [clone: P421]
Type: Primary
Antigen: TTC32
Clonality: Monoclonal
Clone: P421
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR