Anti-TTLL12 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101661
89308-100EA 582.76 USD
89308-100
Anti-TTLL12 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to AMPK gamma 1 (5'-AMP-activated protein kinase subunit gamma-1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IP: Use at an assay dependent dilution. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TTLL12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR