Anti-UBE2T Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101565
89318-482EA 582.76 USD
89318-482
Anti-UBE2T Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CD11a (integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: UBE2T
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR