Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-USP5 Goat Polyclonal Antibody

Supplier: Genetex

GTX10149
89278-764EA 421.12 USD InStock
89278-764
Anti-USP5 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to MYRIP / SLAC2C

Approx 90kDa band observed in Human Brain (Amygdala) lysates (calculated MW of 95.7kDa according to NP_056275.2). Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: USP5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Dog, Monkey, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty