Anti-VWA5A Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX101452
89308-046EA 448.72 USD
89308-046
Anti-VWA5A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to DYNC1H1 (dynein, cytoplasmic 1, heavy chain 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: VWA5A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR