Anti-VWA5A Goat Polyclonal Antibody

Supplier: GeneTex
GTX10145
89287-838EA 607.87 USD
89287-838
Anti-VWA5A Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Pallidin

Approx 23-24kDa band observed in lysates of cell line A431 (calculated MW of 19.7kDa according to NP_036520.1). In transfected HEK293 transiently expressing PLDN a band of approx. 28kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: VWA5A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR