Anti-DISP1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX10142
89278-762EA 602.26 USD
89278-762
Anti-DISP1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Dispatched homolog 1

Preliminary experiments gave no signal but low background in Human Lung and A549 extracts at up to 1µg/ml. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates?

Type: Primary
Antigen: DISP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR