Anti-TRIM5 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX101411
89308-030EA 394.91 USD
89308-030
Anti-TRIM5 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RPS27A (ribosomal protein S27a)

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TRIM5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR