Anti-TBX5 Rabbit Polyclonal Antibody

GTX101375
89318-420EA 376.76 USD
89318-420
Anti-TBX5 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to Alcohol dehydrogenase 1C (alcohol dehydrogenase 1C (class I), gamma polypeptide)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TBX5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR