Anti-PAK2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101372
89318-418EA 582.76 USD
89318-418
Anti-PAK2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Adenosine kinase (adenosine kinase)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PAK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR