Anti-CD40 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX101335
89318-412EA 394.91 USD
89318-412
Anti-CD40 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ASE (arylsulfatase E (chondrodysplasia punctata 1))

  • Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CD40
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR