Anti-TUBB3 Rabbit Polyclonal Antibody

GTX101290
89308-004EA 377.23 USD
89308-004
Anti-TUBB3 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to PAH (phenylalanine hydroxylase)

  • Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: TUBB3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR