Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TUG Mouse Monoclonal Antibody [clone: RS6]

Supplier: Genetex

GTX10128
89336-486EA 246.49 USD InStock
89336-486
Anti-TUG Mouse Monoclonal Antibody [clone: RS6]
Antibodies
Type: Primary
Antigen: TUG
Clonality: Monoclonal
Clone: RS6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty