Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PDE5A Mouse Monoclonal Antibody [clone: RS30]

Supplier: Genetex

GTX10126
89336-484EA 246.49 USD InStock
89336-484
Anti-PDE5A Mouse Monoclonal Antibody [clone: RS30]
Antibodies
Type: Primary
Antigen: PDE5A
Clonality: Monoclonal
Clone: RS30
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty