Anti-CAP-18 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101259
89318-390EA 376.43 USD
89318-390
Anti-CAP-18 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to ELK1 (ELK1, member of ETS oncogene family)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CAP-18
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR