Anti-FLT1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101127
89318-366EA 633.81 USD
89318-366
Anti-FLT1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FLT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR