Anti-MATK Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX101116
89318-360EA 427.1 USD
89318-360
Anti-MATK Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MATK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR