Anti-CER1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101111
89307-968EA 176.03 USD
89307-968
Anti-CER1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: CER1
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR