Anti-ENOS Mouse Monoclonal Antibody [clone: 2B8]

Supplier: GeneTex
GTX10110
89336-482EA 759 USD
89336-482
Anti-ENOS Mouse Monoclonal Antibody [clone: 2B8]
Antibodies
Mouse monoclonal [2B8] to Lactoferrin

ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Detects a band of approximately 77 kDa. Can be used in an ELISA pair with GTX10108 and 10109 as conjugates. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ENOS
Clonality: Monoclonal
Clone: 2B8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR